2-Bromo-6,9-diphenyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A1214188-1G A1214188-250MG A1214188-5G
76678-192EA 49 USD
76678-192 76678-194 77866-698
2-Bromo-6,9-diphenyl-9H-carbazole 98%
2-Bromo-6,9-diphenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆BrN
CAS Number: 1644466-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR