4,4,5,5-Tetramethyl-2-(spiro[fluorene-9,9'-xanthen]-4-yl)-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1214186-1G A1214186-250MG A1214186-5G
76678-186EA 38.2 USD
76678-186 76678-188 76678-190
4,4,5,5-Tetramethyl-2-(spiro[fluorene-9,9'-xanthen]-4-yl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(spiro[fluorene-9,9'-xanthen]-4-yl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₃₁H₂₇BO₃
MW: 458.3553 g/mol
CAS Number: 1609484-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR