Bis(4-(naphthalen-1-yl)phenyl)amine 98%

Supplier: Ambeed
Warning

A1214158-1G A1214158-250MG A1214158-5G
76678-174EA 11 USD
76678-174 76678-176 76678-178
Bis(4-(naphthalen-1-yl)phenyl)amine 98%
Bis(4-(naphthalen-1-yl)phenyl)amine

Precaución: for Research Use Only

Formula: C₃₂H₂₃N
MDL Number: MFCD29765338
CAS Number: 897671-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR