N-([1,1'-Biphenyl]-4-yl)-2'-(9H-carbazol-9-yl)-[1,1'-biphenyl]-4-amine 98%

Supplier: Ambeed
Warning

A1214078-1G A1214078-250MG
76678-168EA 78.51 USD
76678-168 76678-170
N-([1,1'-Biphenyl]-4-yl)-2'-(9H-carbazol-9-yl)-[1,1'-biphenyl]-4-amine 98%
N-([1,1'-Biphenyl]-4-yl)-2'-(9H-carbazol-9-yl)-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₃₆H₂₆N₂
MW: 486.6051 g/mol
CAS Number: 2089223-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR