Ethyl (R)-2-(pyrrolidin-3-yl)acetate hydrochloride 98%

Supplier: Ambeed
Warning

A1213944-1G A1213944-250MG
76678-162EA 110.34 USD
76678-162 76678-164
Ethyl (R)-2-(pyrrolidin-3-yl)acetate hydrochloride 98%
Ethyl (R)-2-(pyrrolidin-3-yl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO₂
MW: 193.6711 g/mol
MDL Number: MFCD24385035
CAS Number: 1332459-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR