(6-(2-Methoxy-2-oxoethyl)pyridin-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

A1213893-1G A1213893-250MG
76678-158EA 330 USD
76678-158 76678-160
(6-(2-Methoxy-2-oxoethyl)pyridin-3-yl)boronic acid 98%
(6-(2-Methoxy-2-oxoethyl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₄
CAS Number: 1620319-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR