N'-Phenyl-3-(2-phenylhydrazono)butanehydrazide 97%

Supplier: Ambeed
Warning

A1213786-100MG A1213786-250MG
76678-154EA 21 USD
76678-154 76678-156
N'-Phenyl-3-(2-phenylhydrazono)butanehydrazide 97%
N'-Phenyl-3-(2-phenylhydrazono)butanehydrazide

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₄O
CAS Number: 67790-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR