6-Bromothieno[3,2-b]pyridin-7-ol 97%

Supplier: Ambeed
Warning

A1213723-100MG A1213723-1G A1213723-250MG
76678-148EA 83.82 USD
76678-148 76678-150 76678-152
6-Bromothieno[3,2-b]pyridin-7-ol 97%
6-Bromothieno[3,2-b]pyridin-7-ol

Precaución: for Research Use Only

Formula: C₇H₄BrNOS
MW: 230.0818 g/mol
MDL Number: MFCD28139683
CAS Number: 875340-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR