(E)-1-(Pyridin-2-yl)ethanone O-benzyl oxime 97%

Supplier: Ambeed
Danger

A1213643-100MG A1213643-250MG
76678-142EA 59.42 USD
76678-142 76678-144
(E)-1-(Pyridin-2-yl)ethanone O-benzyl oxime 97%
(E)-1-(Pyridin-2-yl)ethanone O-benzyl oxime

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O
MW: 226.2738 g/mol
CAS Number: 216753-06-9
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR