Ethyl 2,4,5-trichlorobenzoate 98%

Supplier: Ambeed
Warning

A1213555-100G A1213555-25G A1213555-5G
76678-136EA 172 USD
76678-136 76678-138 76678-140
Ethyl 2,4,5-trichlorobenzoate 98%
Ethyl 2,4,5-trichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇Cl₃O₂
MDL Number: MFCD02149442
CAS Number: 86569-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR