2-(Methylamino)ethan-1-ol hydrochloride 95%

Supplier: Ambeed
Warning

A1213382-1G A1213382-250MG
76678-122EA 305 USD
76678-122 77571-268
2-(Methylamino)ethan-1-ol hydrochloride 95%
2-(Methylamino)ethan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₃H₁₀ClNO
CAS Number: 62640-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR