(E)-Cyclooct-2-en-1-yl (4-nitrophenyl) carbonate 95% rel-(1R-2E-pS)-axial

Supplier: Ambeed
Warning

A1213336-50MG A1213336-10MG A1213336-250MG A1213336-25MG
76678-118EA 395.72 USD
76678-118 77620-916 77620-918 77620-920
(E)-Cyclooct-2-en-1-yl (4-nitrophenyl) carbonate 95% rel-(1R-2E-pS)-axial
(E)-Cyclooct-2-en-1-yl (4-nitrophenyl) carbonate

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₅
MW: 291.2992 g/mol
CAS Number: 1580501-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR