4-Methoxybenzyl mercaptan 96%

Supplier: Ambeed
Danger

A1213240-1G A1213240-25G A1213240-5G A1213240-100G
76678-092EA 16 USD
76678-092 76678-094 77549-424 77620-906
4-Methoxybenzyl mercaptan 96%
4-Methoxybenzyl mercaptan

Precaución: for Research Use Only

Formula: C₈H₁₀OS
MDL Number: MFCD00004871
CAS Number: 6258-60-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR