4,5-Dibromo-3-methylisothiazole 97%

Supplier: Ambeed
Danger

A1212424-100MG A1212424-250MG
76678-076EA 10 USD
76678-076 77866-681
4,5-Dibromo-3-methylisothiazole 97%
4,5-Dibromo-3-methylisothiazole

Precaución: for Research Use Only

Formula: C₄H₃Br₂NS
MW: 256.9463 g/mol
MDL Number: MFCD00233954
CAS Number: 1732-55-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR