Methyl 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate 98%

Supplier: Ambeed
Warning

A1212383-1G A1212383-250MG A1212383-5G
76678-062EA 24.41 USD
76678-062 76678-064 76678-066
Methyl 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate 98%
Methyl 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇BO₄
MW: 248.0827 g/mol
CAS Number: 849412-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR