6-(Ethyl(3-isopropoxy-4-isopropylphenyl)amino)nicotinic acid 95%

Supplier: Ambeed
Warning

A1212308-100MG A1212308-250MG
76678-058EA 194.15 USD
76678-058 76678-060
6-(Ethyl(3-isopropoxy-4-isopropylphenyl)amino)nicotinic acid 95%
6-(Ethyl(3-isopropoxy-4-isopropylphenyl)amino)nicotinic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₆N₂O₃
MW: 342.4321 g/mol
CAS Number: 1041186-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR