Methyl (S)-2-(chloromethyl)-1-(oxetan-2-ylmethyl)-1H-benzo[d]imidazole-6-carboxylate 98%

Supplier: Ambeed
Danger

A1212083-100MG A1212083-250MG A1212083-10G A1212083-1G A1212083-5G A1212083-25G
76678-052EA 12 USD
76678-052 76678-054 77620-886 77620-888 77866-680 77866-679
Methyl (S)-2-(chloromethyl)-1-(oxetan-2-ylmethyl)-1H-benzo[d]imidazole-6-carboxylate 98%
Methyl (S)-2-(chloromethyl)-1-(oxetan-2-ylmethyl)-1H-benzo[d]imidazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₅ClN₂O₃
MDL Number: MFCD32068480
CAS Number: 2230200-76-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR