Methyl 2-(3,5-dibromo-4-cyanophenyl)acetate 95%

Supplier: Ambeed
Warning

A1211894-1G A1211894-250MG
76678-048EA 301.3 USD
76678-048 76678-050
Methyl 2-(3,5-dibromo-4-cyanophenyl)acetate 95%
Methyl 2-(3,5-dibromo-4-cyanophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₇Br₂NO₂
MW: 332.9761 g/mol
MDL Number: MFCD28758583
CAS Number: 1804892-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR