2,4-Dibromo-3-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A1211765-1G A1211765-250MG
76678-038EA 84 USD
76678-038 77866-675
2,4-Dibromo-3-hydroxybenzoic acid 98%
2,4-Dibromo-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Br₂O₃
MDL Number: MFCD32205364
CAS Number: 91659-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR