5-Chloro-4-hydroxy-2-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A1211753-1G A1211753-250MG A1211753-5G
76678-028EA 120.95 USD
76678-028 76678-030 76678-032
5-Chloro-4-hydroxy-2-methylbenzaldehyde 97%
5-Chloro-4-hydroxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.5930 g/mol
MDL Number: MFCD22414661
CAS Number: 1556030-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR