Methyl 2-oxo-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

A1211675-100MG A1211675-1G A1211675-250MG A1211675-5G
76678-016EA 58.35 USD
76678-016 76678-018 76678-020 76678-022
Methyl 2-oxo-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-3-carboxylate 97%
Methyl 2-oxo-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MW: 192.1714 g/mol
MDL Number: MFCD21294394
CAS Number: 1356542-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR