4-((4-Methylpiperazin-1-yl)methyl)benzoyl chloride hydrochloride 97%

Supplier: Ambeed
Danger

A1211493-100MG A1211493-1G A1211493-250MG A1211493-25G A1211493-5G
76678-006EA 6.37 USD
76678-006 76678-008 76678-010 76678-012 76678-014
4-((4-Methylpiperazin-1-yl)methyl)benzoyl chloride hydrochloride 97%
4-((4-Methylpiperazin-1-yl)methyl)benzoyl chloride hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₈Cl₂N₂O
MW: 289.2008 g/mol
CAS Number: 909252-77-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR