4-Amino-5-(trifluoromethyl)isophthalonitrile 97%

Supplier: Ambeed
Danger

A1211446-1G A1211446-250MG
76677-998EA 98 USD
76677-998 76678-000
4-Amino-5-(trifluoromethyl)isophthalonitrile 97%
4-Amino-5-(trifluoromethyl)isophthalonitrile

Precaución: for Research Use Only

Formula: C₉H₄F₃N₃
CAS Number: 2442597-62-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR