Boc-3-Cyclopropylalanine 98%

Supplier: Ambeed
Warning

Synonyms: 2-((tert-Butoxycarbonyl)amino)-3-cyclopropylpropanoic acid

A1211067-100MG A1211067-1G A1211067-250MG
76677-986EA 30.77 USD
76677-986 76677-988 76677-990
Boc-3-Cyclopropylalanine 98%
Boc-3-Cyclopropylalanine

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.273 g/mol
MDL Number: MFCD08061239
CAS Number: 888323-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR