6-[4-(3-Chlorophenyl)piperazin-1-yl]-3-cyclohexyl-1,2,3,4-tetrahydropyrimidine-2,4-dione 98+%

Supplier: Ambeed
Warning

A1210813-10MG A1210813-1MG A1210813-50MG A1210813-5MG A1210813-250MG
76677-978EA 58 USD
76677-978 76677-980 76677-982 76677-984 77866-663
6-[4-(3-Chlorophenyl)piperazin-1-yl]-3-cyclohexyl-1,2,3,4-tetrahydropyrimidine-2,4-dione 98+%
6-[4-(3-Chlorophenyl)piperazin-1-yl]-3-cyclohexyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

Precaución: for Research Use Only

Formula: C₂₀H₂₅ClN₄O₂
MDL Number: MFCD05894342
CAS Number: 863588-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR