8-((1H-Imidazol-4-yl)methylene)-6,8-dihydro-7H-thiazolo[5,4-e]indol-7-one 98+%

Supplier: Ambeed
Warning

Synonyms: PKR-IN-C16

A1209816-10MG A1209816-1MG A1209816-5MG
76677-956EA 114.58 USD
76677-956 76677-958 76677-960
8-((1H-Imidazol-4-yl)methylene)-6,8-dihydro-7H-thiazolo[5,4-e]indol-7-one 98+%
8-((1H-Imidazol-4-yl)methylene)-6,8-dihydro-7H-thiazolo[5,4-e]indol-7-one

Precaución: for Research Use Only

Formula: C₁₃H₈N₄OS
MW: 268.2938 g/mol
MDL Number: MFCD28046009
CAS Number: 608512-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR