2-(Adamantan-1-yl)-N-(2-((2-((2-hydroxyethyl)amino)ethyl)amino)quinolin-5-yl)acetamide dihydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: AZ10606120 2HCl

A1209771-100MG A1209771-5MG A1209771-50MG
76677-954EA 151 USD
76677-954 77620-862 77866-649
2-(Adamantan-1-yl)-N-(2-((2-((2-hydroxyethyl)amino)ethyl)amino)quinolin-5-yl)acetamide dihydrochloride 98+%
2-(Adamantan-1-yl)-N-(2-((2-((2-hydroxyethyl)amino)ethyl)amino)quinolin-5-yl)acetamide dihydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₃₆Cl₂N₄O₂
MDL Number: MFCD12828760
CAS Number: 607378-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR