FBPase-1 inhibitor ≥98%

Supplier: Ambeed
Warning

A1209735-100MG A1209735-50MG
76677-952EA 130 USD
76677-952 77620-860
FBPase-1 inhibitor ≥98%
FBPase-1 inhibitor

Precaución: for Research Use Only

Formula: C₁₃H₇Cl₃N₂O₃S
MDL Number: MFCD22581250
CAS Number: 883973-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR