rel-2-(4-Chlorophenoxy)-2-methyl-N-((1S,2R)-5-(methylsulfonamido)adamantan-2-yl)propanamide 98%

Supplier: Ambeed
Warning

Synonyms: JNJ 303

A1209078-10MG A1209078-1MG A1209078-50MG A1209078-5MG
76677-916EA 68.96 USD
76677-916 76677-918 76677-920 76677-922
rel-2-(4-Chlorophenoxy)-2-methyl-N-((1S,2R)-5-(methylsulfonamido)adamantan-2-yl)propanamide 98%
rel-2-(4-Chlorophenoxy)-2-methyl-N-((1S,2R)-5-(methylsulfonamido)adamantan-2-yl)propanamide

Precaución: for Research Use Only

Formula: C₂₁H₂₉ClN₂O₄S
MW: 440.98 g/mol
MDL Number: MFCD18086905
CAS Number: 878489-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR