(3,4,5-Trimethoxybenzyl)hydrazine hydrochloride 98%

Supplier: Ambeed
Warning

A1208762-1G A1208762-250MG
76677-910EA 86 USD
76677-910 76677-912
(3,4,5-Trimethoxybenzyl)hydrazine hydrochloride 98%
(3,4,5-Trimethoxybenzyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₇ClN₂O₃
MDL Number: MFCD12031974
CAS Number: 3903-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR