L(+)-Sodium lactate 98% (contains SiO2)

Supplier: Ambeed
Warning

A1208667-1000G A1208667-100G A1208667-25G A1208667-500G
76677-902EA 34 USD
76677-902 76677-904 76677-906 76677-908
L(+)-Sodium lactate 98% (contains SiO2)
L(+)-Sodium lactate

Precaución: for Research Use Only

Formula: C₃H₅NaO₃
MDL Number: MFCD00066576
CAS Number: 867-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR