(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl propionate 97%

Supplier: Ambeed
Warning

Synonyms: MIRA-1

A1208571-1G A1208571-250MG A1208571-5G A1208571-100MG
76677-896EA 220.67 USD
76677-896 76677-898 76677-900 77596-268
(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl propionate 97%
(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl propionate

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MW: 183.1614 g/mol
MDL Number: MFCD12828765
CAS Number: 72835-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR