4-Hydroxyphenyl methacrylate 97%

Supplier: Ambeed
Warning

A1208549-25G A1208549-5G
76677-894EA 43 USD
76677-894 77866-628
4-Hydroxyphenyl methacrylate 97%
4-Hydroxyphenyl methacrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.1846 g/mol
MDL Number: MFCD00227728
CAS Number: 31480-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR