(R)-1-(5-Bromopyridin-3-yl)ethanamine 97%

Supplier: Ambeed
Warning

A1208037-1G A1208037-250MG A1208037-100MG
76677-876EA 612.14 USD
76677-876 76677-878 77596-262
(R)-1-(5-Bromopyridin-3-yl)ethanamine 97%
(R)-1-(5-Bromopyridin-3-yl)ethanamine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MW: 201.0638 g/mol
MDL Number: MFCD09256750
CAS Number: 1212905-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR