2-(Naphthalen-2-yl)ethanamine hydrobromide 97%

Supplier: Ambeed
Danger

A1207693-1G A1207693-250MG A1207693-5G
76677-846EA 32.89 USD
76677-846 76677-848 76677-850
2-(Naphthalen-2-yl)ethanamine hydrobromide 97%
2-(Naphthalen-2-yl)ethanamine hydrobromide

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrN
MW: 252.1503 g/mol
MDL Number: MFCD33022355
CAS Number: 2668984-44-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR