N-((1S,2S)-2-Amino-1,2-bis(4-methoxyphenyl)ethyl)-4-methylbenzenesulfonamide 97%

Supplier: Ambeed
Warning

A1207658-100MG A1207658-1G A1207658-250MG
76677-814EA 15.92 USD
76677-814 76677-816 76677-818
N-((1S,2S)-2-Amino-1,2-bis(4-methoxyphenyl)ethyl)-4-methylbenzenesulfonamide 97%
N-((1S,2S)-2-Amino-1,2-bis(4-methoxyphenyl)ethyl)-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₄S
MW: 426.5285 g/mol
CAS Number: 887924-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR