N-((1R,2R)-2-Amino-1,2-bis(4-methoxyphenyl)ethyl)-4-methylbenzenesulfonamide 97%

Supplier: Ambeed
Warning

A1207657-100MG A1207657-1G A1207657-250MG
76677-808EA 21.22 USD
76677-808 76677-810 76677-812
N-((1R,2R)-2-Amino-1,2-bis(4-methoxyphenyl)ethyl)-4-methylbenzenesulfonamide 97%
N-((1R,2R)-2-Amino-1,2-bis(4-methoxyphenyl)ethyl)-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₄S
MW: 426.5285 g/mol
CAS Number: 852212-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR