(S)-3-(tert-Butyl)-4-(2-methoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 95%

Supplier: Ambeed
Warning

A1207639-100MG A1207639-1G
76677-784EA 74 USD
76677-784 77866-609
(S)-3-(tert-Butyl)-4-(2-methoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 95%
(S)-3-(tert-Butyl)-4-(2-methoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

Precaución: for Research Use Only

Formula: C₁₈H₂₁O₂P
MDL Number: MFCD32641178
CAS Number: 2448646-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR