2-(3-Fluoropyridin-4-yl)propan-2-amine hydrochloride 97%

Supplier: Ambeed
Warning

A1207415-100MG A1207415-250MG
76677-750EA 135 USD
76677-750 76677-752
2-(3-Fluoropyridin-4-yl)propan-2-amine hydrochloride 97%
2-(3-Fluoropyridin-4-yl)propan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClFN₂
CAS Number: 2442597-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR