1-(3-Fluorophenyl)-5-methylpyridin-2(1H)-one 98%
Supplier: Ambeed
|
Warning
|
A1207399-10MG
A1207399-1MG
A1207399-25MG
A1207399-50MG
A1207399-5MG
A1207399-100MG
A1207399-250MG
76677-740EA
43
USD
76677-740
76677-742
76677-744
76677-746
76677-748
77866-594
77866-595
1-(3-Fluorophenyl)-5-methylpyridin-2(1H)-one 98%
1-(3-Fluorophenyl)-5-methylpyridin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₁₂H₁₀FNO |
MDL Number:
MFCD29088959 CAS Number: 848353-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...