1-Iminothiomorpholine 1-oxide hydrochloride 95%

Supplier: Ambeed
Warning

A1207382-100MG A1207382-250MG A1207382-1G A1207382-50MG
76677-734EA 146 USD
76677-734 77571-236 77620-816 77866-592
1-Iminothiomorpholine 1-oxide hydrochloride 95%
1-Iminothiomorpholine 1-oxide hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClN₂OS
MDL Number: MFCD30833937
CAS Number: 1633667-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR