3,6-Di-tert-butyl-9-mesityl-10-methylacridin-10-ium tetrafluoroborate 97%

Supplier: Ambeed
Warning

A1207362-100MG A1207362-1G A1207362-250MG A1207362-50MG
76677-724EA 58.35 USD
76677-724 76677-726 76677-728 76677-730
3,6-Di-tert-butyl-9-mesityl-10-methylacridin-10-ium tetrafluoroborate 97%
3,6-Di-tert-butyl-9-mesityl-10-methylacridin-10-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₃₁H₃₈BF₄N
MW: 511.4447 g/mol
CAS Number: 2054779-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR