9-Mesityl-2,7,10-trimethylacridin-10-ium tetrafluoroborate 97%
Supplier: Ambeed
|
Warning
|
A1207360-100MG
A1207360-250MG
76677-720EA
127.31
USD
76677-720
76677-722
9-Mesityl-2,7,10-trimethylacridin-10-ium tetrafluoroborate 97%
9-Mesityl-2,7,10-trimethylacridin-10-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₅H₂₆BF₄N MW: 427.2853 g/mol |
MDL Number:
MFCD32173817 CAS Number: 1621019-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...