9-Mesityl-10-methylacridin-10-ium hexafluorophosphate(V) 97%

Supplier: Ambeed
Warning

A1207356-100MG A1207356-1G A1207356-250MG
76677-714EA 48.81 USD
76677-714 76677-716 76677-718
9-Mesityl-10-methylacridin-10-ium hexafluorophosphate(V) 97%
9-Mesityl-10-methylacridin-10-ium hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₂₃H₂₂F₆NP
MW: 457.3917 g/mol
CAS Number: 872205-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR