9-Mesityl-2,7-dimethoxy-10-phenylacridin-10-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1207350-100MG A1207350-250MG
76677-710EA 58.35 USD
76677-710 76677-712
9-Mesityl-2,7-dimethoxy-10-phenylacridin-10-ium tetrafluoroborate 97%
9-Mesityl-2,7-dimethoxy-10-phenylacridin-10-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₃₀H₂₈BF₄NO₂
MW: 521.3534 g/mol
CAS Number: 1965330-55-5
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR