Hexyl 2-methoxyacetate 97%

Supplier: Ambeed
Warning

A1207334-1G A1207334-250MG A1207334-5G
76677-704EA 26.53 USD
76677-704 76677-706 76677-708
Hexyl 2-methoxyacetate 97%
Hexyl 2-methoxyacetate

Precaución: for Research Use Only

Formula: C₉H₁₈O₃
MW: 174.2374 g/mol
MDL Number: MFCD32644594
CAS Number: 145747-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR