Heptyl hexanoate 97%

Supplier: Ambeed
Warning

A1207332-100MG A1207332-1G A1207332-250MG
76677-698EA 42.44 USD
76677-698 76677-700 76677-702
Heptyl hexanoate 97%
Heptyl hexanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₆O₂
MW: 214.3443 g/mol
MDL Number: MFCD00048909
CAS Number: 6976-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR