8-Bromo-3-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A1207322-100MG A1207322-1G A1207322-250MG
76677-692EA 23 USD
76677-692 76677-694 76677-696
8-Bromo-3-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine 97%
8-Bromo-3-iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₃BrF₃IN₂
MW: 390.9265 g/mol
MDL Number: MFCD30834260
CAS Number: 2384795-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR