1-Bromo-4-(perfluorohexyl)benzene 97%

Supplier: Ambeed
Warning

A1207314-1G A1207314-250MG A1207314-5G
76677-686EA 546 USD
76677-686 76677-688 76677-690
1-Bromo-4-(perfluorohexyl)benzene 97%
1-Bromo-4-(perfluorohexyl)benzene

Precaución: for Research Use Only

Formula: C₁₂H₄BrF₁₃
MDL Number: MFCD03788327
CAS Number: 149068-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR