4-Chloro-1-methyl-3-nitropyridin-2(1H)-one 97%

Supplier: Ambeed
Danger

A1207288-1G A1207288-250MG
76677-682EA 318 USD
76677-682 76677-684
4-Chloro-1-methyl-3-nitropyridin-2(1H)-one 97%
4-Chloro-1-methyl-3-nitropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₃
MDL Number: MFCD25949141
CAS Number: 719268-89-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR